missing translation for 'onlineSavingsMsg'
Learn More

Perylene (purified by sublimation) 99.0+%, TCI America™

Catalog No. P16291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16291G Supplier TCI America Supplier No. P16291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 198-55-0
Molecular Formula C20H12
Molecular Weight (g/mol) 252.316
MDL Number MFCD00004142
InChI Key CSHWQDPOILHKBI-UHFFFAOYSA-N
Synonym peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl
PubChem CID 9142
ChEBI CHEBI:29861
IUPAC Name perylene
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2

Specifications

Melting Point 278°C
Quantity 1 g
Formula Weight 252.32
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Perylene (purified by sublimation)
RTECSNumber SE3794000
TSCA Yes