missing translation for 'onlineSavingsMsg'
Learn More

Perylene, 98+%

Catalog No. AAL0304703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0304703 1 g
AAL0304706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0304703 Supplier Thermo Scientific Chemicals Supplier No. L0304703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 198-55-0 | C20H12 | 252.316 g/mol

Perylene is used as a fluorescent and a chemiluminescent agent in the cytochemistry of membranes. It serves as a blue-emitting dopant material in organic light emitting diodes. It finds application as an organic photoconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perylene is used as a fluorescent and a chemiluminescent agent in the cytochemistry of membranes. It serves as a blue-emitting dopant material in organic light emitting diodes. It finds application as an organic photoconductor.

Solubility
Soluble in chloroform, carbon disulfide and benzene. Slightly soluble in alcohol and ethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 198-55-0
Molecular Formula C20H12
Molecular Weight (g/mol) 252.316
MDL Number MFCD00004142
InChI Key CSHWQDPOILHKBI-UHFFFAOYSA-N
Synonym peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl
PubChem CID 9142
ChEBI CHEBI:29861
IUPAC Name perylene
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2

Specifications

Melting Point 276°C to 280°C
Odor Odorless
Quantity 1 g
Beilstein 1911335
Merck Index 14,7184
Solubility Information Soluble in chloroform,carbon disulfide and benzene. Slightly soluble in alcohol and ethyl ether. Insoluble in water.
Formula Weight 252.32
Percent Purity ≥98%
Chemical Name or Material Perylene
EINECSNumber 205-900-9
RTECSNumber SE3794000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only