missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water

CAS: 13768-11-1 | HO4Re | 251.21 g/mol

$240.21 - $794.35

Chemical Identifiers

CAS 13768-11-1
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
Synonym perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
PubChem CID 83718
IUPAC Name hydroxy(trioxo)rhenium
SMILES O[Re](=O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC316180050
SDS View Documents
Thermo Scientific Chemicals
316180050
5 g Glass bottle
Each for $240.21
Only null left
 
AC316180250
SDS View Documents
Thermo Scientific Chemicals
316180250
25 g Glass bottle
Each for $794.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Reduction reagent
Specifications

Chemical Identifiers

13768-11-1
251.21
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
83718
O[Re](=O)(=O)=O
HO4Re
MFCD00011326
perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
hydroxy(trioxo)rhenium

Specifications

Colorless to Yellow
99.99%
HReO4
70 to 76.5% (HReO4)
perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
O[Re](=O)(=O)=O
251.21
251.21
Glass bottle
Solution
13768-11-1
HO4Re
MFCD00011326
2.1600g/mL
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
hydroxy(trioxo)rhenium
83718
Trace Metal Basis
5 g
Perrhenic(VII)acid, 99.99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

RUO – Research Use Only