missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water

Catalog No. AC316180250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC316180250 25 g Glass bottle
AC316180050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC316180250 Supplier Thermo Scientific Chemicals Supplier No. 316180250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13768-11-1 | HO4Re | 251.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Reduction reagent

Chemical Identifiers

CAS 13768-11-1
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
Synonym perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
PubChem CID 83718
IUPAC Name hydroxy(trioxo)rhenium
SMILES O[Re](=O)(=O)=O

Specifications

Color Colorless to Yellow
Concentration 70 to 76.5% (HReO4)
Density 2.1600g/mL
Packaging Glass bottle
Assay Percent Range 99.99%
Linear Formula HReO4
Quantity 25 g
Physical Form Solution
Formula Weight 251.21
Grade Trace Metal Basis
Chemical Name or Material Perrhenic(VII)acid, 99.99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only