missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
Volume ranges from 20 to 200μL
$877.91 - $5630.65
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| For Use With (Equipment) | PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head |
| Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Color | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Color | Price | Quantity | |||
|
14-222-442
|
Axygen™
RRF175CRS |
175 μL | Sterile | Clear |
Case of 4800 Each for $4,218.17
|
|
|||
|
14-222-448
|
Axygen™
RRP200CRS |
200 μL | Sterile | Clear |
Case of 4800 Each for $4,074.57
|
|
|||
|
14-222-443
|
Axygen™
RRF25CBKRS |
25 μL | Sterile | Black |
Pack of 960 Each for $877.91
Case of 5 Pack for $4,389.55
|
|
|||
|
14-222-453
|
Axygen™
RRP50CBKR |
50 μL | Non-sterile | Black |
Case of 4800 Each for $4,255.22
|
|
|||
|
14-222-445
|
Axygen™
RRP200CBKR |
200 μL | Non-sterile | Black |
Case of 4800 Each for $4,514.96
|
|
|||
|
14-222-444
|
Axygen™
RRF25CRS |
25 μL | Sterile | Clear |
Case of 4800 Each for $4,218.17
|
|
|||
|
14-222-441
|
Axygen™
RRF175CBKRS |
175 μL | Sterile | Black |
Case of 4800 Each for $5,630.65
|
|
|||
|
14-222-447
|
Axygen™
RRP200CR |
200 μL | Non-sterile | Clear |
Case of 4800 Each for $3,951.74
|
|
|||
|
14-222-451
|
Axygen™
RRP20CR |
20 μL | Non-sterile | Clear |
Case of 4800 Each for $3,623.48
|
|
|||
Description
- Racked
- Black and Clear tips available
- With Liquid level sensing
- Nonsterile and Presterilized tips
- Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
| Autoclavable | |
| DNase, RNase, and Pyrogen free | |
| Rack | |
| PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head |