missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips

Catalog No. 14222453
Change view
Click to view available options
Volume (Metric):
20 μL
25 μL
50 μL
175 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Clear
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
14222453 50 μL Non-sterile Black
14222442 175 μL Sterile Clear
14222448 200 μL Sterile Clear
14222443 25 μL Sterile Black
14222445 200 μL Non-sterile Black
14222444 25 μL Sterile Clear
14222441 175 μL Sterile Black
14222447 200 μL Non-sterile Clear
14222451 20 μL Non-sterile Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14222453 Supplier Axygen™ Supplier No. RRP50CBKR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Volume ranges from 20 to 200μL

  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Rack Color Green
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Liquid Level Sensing, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head
Material Polypropylene
Show More Show Less