Learn More
Agilent BioTek Peristaltic Pump Dispenser Cassette with Large Bore Plastic Tips, 5 μL Increments
Catalog No. BT7170024
Change view
Click to view available options
For Use With (Equipment):
EL406 Microplate Washer, 406 FX Microplate Washer
Catalog No. For Use With (Equipment)
BT7170024 EL406 Microplate Washer, 406 FX Microplate Washer
1 options
Catalog No. BT7170024 Supplier Agilent BioTek Supplier No. 7170024
Dispenser Cassette with Large Bore Plastic Tips for use with the EL406, 406 FX, MultiFlo and MultiFlo FX.
  • 5 μL increments
Specifications
Capacity (Metric) 5 to 2500 μL
Type Cassette
For Use With (Equipment) EL406 Microplate Washer, 406 FX Microplate Washer