missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Periostin (Postn) ELISA Kit

Catalog No. EMPOSTN
Encompass
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMPOSTN 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EMPOSTN Supplier Invitrogen™ Supplier No. EMPOSTN

ELISA

The Mouse Periostin (Postn) ELISA quantitates Ms Periostin in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Periostin. Principle of the method The Mouse Periostin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Periostin (PN), also designated osteoblast-specific factor 2 (OSF-2), is a disulfide linked protein originally isolated as a osteoblast-specific factor. Periostin is a secreted protein that binds heparin and functions as a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins. In preosteoblasts, Periostin acts as a cell adhesion molecule and plays a role in osteoblast recruitment, spreading and attachment. Periostin is mainly detected in lower gastrointestinal tract, aorta, stomach, placenta, uterus and breast tissues but is up-regulated in epithelial ovarian tumors and overexpressed in breast cancer. Expression of Periostin is increased by bone morphogenetic protein (BMP2) and transforming growth factor beta 1(TGF beta 1). Periostin contains a typical signal sequence, followed by a cysteine-rich domain, a fourfold repeated domain, which shows homology with the insect protein fasciclin, and a C-terminal domain.

Specifications

Accession Number Q62009
Assay Range 0.66 to 16 ng/mL
Assay Sensitivity 0.05 ng/mL
Calibration Range 0.05 ng/mL to 16 ng/mL
Description Periostin (Postn) ELISA Kit, 96 Assays, 1 Kit, Mouse Species, HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of Periostin, Osteoblast Specific Factor (Postn) in Samples Using a M
Detectable Analytes Periostin, osteoblast specific factor (Postn)
Assay Solid-phase sandwich ELISA
Conjugate HRP
Detection Method Colorimetric
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: Axl, BLC, CD27 Ligand, CD30, CD30 Ligand, CD40, CXCL16, EGF, E-selectin, Fractalkine, GITR, HGF, ICAM-1, IFN-gamma, IGF-1, IGFBP-2, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 ra, IL-1 beta, IL-2, IL-2 R alpha, IL-3, IL-4, IL-5, IL-6, IL-7, IL-10, IL-12 p70, IL-17 E, IL-17 F, IL-20, IL-23, IL-28, I-TAC, MCP-1, MCP-5, M-CSF, MDC, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 alpha, Osteopontin, Osteoprotegerin, Prolactin, Pro-MMP-9, P-selectin, Resistin, SCF, SDF-1 alpha, TPO, VCAM-1.
Sufficient For 96 Assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 50706
Gene Symbol A630052E07Rik,AI747096,OSF-2,Osf2,PLF,PN
Host Species Mouse
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 2 μL, Serum 2 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym A630052E07Rik, AI747096, OSF-2, Osf2, PLF, PN, peri
Target Periostin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less