missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, for analysis

Catalog No. p-4813598
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453170250 25 g
AC453171000 100 g
AC453175000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC453170250 Supplier Thermo Scientific Chemicals Supplier No. 453170250
Only null left

CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
SMILES O[I](O)(O)(O)(O)=O

Specifications

Melting Point 122.0°C
Quantity 25 g
Linear Formula H5IO6
Solubility Information Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol, slightly soluble in ether
Formula Weight 227.94
Grade Analytical
Chemical Name or Material Periodic Acid

RUO – Research Use Only