missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periodic Acid, Reagent Grade, Reagents
Periodic Acid, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C126725025C1
Specifications
| Periodic Acid, Reagent Grade | |
| 10450-60-9 | |
| Glass Bottle | |
| 25 g | |
| Solid | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| tetrahydroxyiodous acid | |
| Reagent |
| White to Pale Yellow | |
| 140°C | |
| 1.4 | |
| H5IO6 | |
| Iodine Tetraoxide | |
| O[I](O)(O)(O)(O)=O | |
| 227.94 |
Chemical Identifiers
| 10450-60-9 | |
| 227.94 | |
| Iodine Tetraoxide | |
| O[I](O)(O)(O)(O)=O |
| H5IO6 | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| tetrahydroxyiodous acid |