missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid (Certified) ACS, Fisher Chemical™

Catalog No. p-32737
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A22325 25 g Glass Bottle
A223100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22325 Supplier Thermo Fisher Scientific Supplier No. FLA22325
Only null left

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

pH 2
Color White-Yellow
Density 1.4 g/cm3
Identification Pass Test
Packaging Glass Bottle
Decomposition Information 122°C
Assay Percent Range ≥99%
Quantity 25 g
Physical Form Solid
UN Number 3085
Formula Weight 227.96
Percent Purity ≥99%
Grade Certified
Chemical Name or Material Periodic Acid
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer