missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, Approx. 98%, Spectrum™ Chemical

Catalog No. 18612191
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612191 100 g Amber Glass Bottle
18612193 500 g Amber Glass Bottle
18612192 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612191 Supplier Spectrum Chemical Mfg Cor Supplier No. P1036100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 10450-60-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity ∼98%
Grade Ungraded
Chemical Name or Material Periodic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.