Learn More
Periodic acid, 34% w/w aq. soln.
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
$264.79 - $264.79
Chemical Identifiers
| CAS | 10450-60-9 |
|---|---|
| Molecular Formula | H5IO6 |
| Molecular Weight (g/mol) | 227.94 |
| MDL Number | MFCD00011344 |
| InChI Key | TWLXDPFBEPBAQB-UHFFFAOYSA-N |
| Synonym | orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst |
| PubChem CID | 25289 |
| ChEBI | CHEBI:29150 |
| IUPAC Name | pentahydroxy(oxo)-$l^{7}-iodane |
| SMILES | O[I](O)(O)(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2017236
|
Thermo Scientific Chemicals
B2017236 |
500 g |
Each for $264.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10450-60-9 | |
| 227.94 | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| 25289 | |
| pentahydroxy(oxo)-$l^{7}-iodane |
| H5IO6 | |
| MFCD00011344 | |
| orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
| CHEBI:29150 | |
| O[I](O)(O)(O)(O)=O |
Specifications
| 10450-60-9 | |
| Light sensitive | |
| H5IO6 | |
| UN3098 | |
| orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
| O[I](O)(O)(O)(O)=O | |
| 227.94 | |
| CHEBI:29150 | |
| Periodic acid |
| 34% w/w aq. soln. | |
| 500 g | |
| MFCD00011344 | |
| 14,7171 | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| pentahydroxy(oxo)-$l^{7}-iodane | |
| 25289 | |
| 227.94 |
Safety and Handling
GHS H Statement
H272-H314
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 233-937-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only