Learn More
Periodic Acid, 0.5%, StatLab™
A ready to use periodic acid solution used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen.
Supplier: Statlab AHP0343
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Description
- A component for the following MasterTech Stain Kits: P.A.S. Stain Kit, Alcian Blue-P.A.S. Stain Kit, and Jone's Basement Membrane Reticulum Stain Kit
Specifications
| Clear | |
| Bottle | |
| 1 pt. |