missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periodic Acid, 0.5%, StatLab™
Description
- A component for the following MasterTech Stain Kits: P.A.S. Stain Kit, Alcian Blue-P.A.S. Stain Kit, and Jone's Basement Membrane Reticulum Stain Kit
Specifications
Specifications
| Color | Clear |
| Packaging | Bottle |
| Quantity | 100 mL |