missing translation for 'onlineSavingsMsg'
Learn More

Perindopril erbumine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012406
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012406 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012406 Supplier Merck Emd Millipore Supplier No. PHR1887500MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 107133-36-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H32N2O5 · C4H11 N
MDL Number MFCD02313824
Synonym Perindopril erbumine; (2 S,3 AS,7 AS)-1-[(2 S)-2-[[(1 S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1 H-indole-2-carboxylic acid tert-butylamine salt; Perindopril tert-butylamine; S-9490
Type Certified Reference Material
Quantity 500 mg
Formula Weight 441.60
Grade Certified Reference Material