missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotri-n-butylamine, tech. 90%, Thermo Scientific Chemicals

Catalog No. p-7024658
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1971206 5 g
AAA1971214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1971206 Supplier Thermo Scientific Chemicals Supplier No. A1971206
Only null left

Perfluorotri-n-butylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorotri-n-butylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.

Solubility
Miscible with chloroform, cyclohexane, ethyl ether, ethyl acetate, heptane, carbon tetrachloride and petroleum ether. Immiscible with water, methanol, toluene, isopropyl alcohol, benzene and acetone.

Notes
Store in a cool place. Incompatible with powdered metals and alkali metals.

Chemical Identifiers

CAS 311-89-7
Molecular Formula C12F27N
Molecular Weight (g/mol) 671.096
MDL Number MFCD00000436
InChI Key RVZRBWKZFJCCIB-UHFFFAOYSA-N
Synonym perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine
PubChem CID 9397
ChEBI CHEBI:38854
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point -52°C
Density 1.88
Boiling Point 177°C to 180°C
Refractive Index 1.289
Quantity 5 g
Beilstein 1813883
Solubility Information Miscible with chloroform,cyclohexane,ethyl ether,ethyl acetate,heptane,carbon tetrachloride and petroleum ether. Immiscible with water,methanol,toluene,isopropyl alcohol,benzene and acetone.
Formula Weight 671.1
Percent Purity 90%
Grade Technical
Chemical Name or Material Perfluorotri-n-butylamine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 206-223-1
RTECSNumber YA1000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only