Learn More
Perfluorotetradecanoic acid, 96%
CAS: 376-06-7 | C14HF27O2 | 714.117 g/mol
$55.36 - $148.33
Chemical Identifiers
| CAS | 376-06-7 |
|---|---|
| Molecular Formula | C14HF27O2 |
| Molecular Weight (g/mol) | 714.117 |
| MDL Number | MFCD00153263 |
| InChI Key | RUDINRUXCKIXAJ-UHFFFAOYSA-N |
| Synonym | perfluorotetradecanoic acid, perfluoromyristic acid, heptacosafluorotetradecanoic acid, tetradecanoic acid, heptacosafluoro, acmc-20ajxq, perfluorotetradecanoic acid, analytical standard |
| PubChem CID | 67822 |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid |
| SMILES | C(=O)(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL1379603
|
N/A | 1 g |
Each for $55.36
|
|
|||||
|
AAL1379606
|
N/A | 5 g |
Each for $148.33
|
|
|||||
Description
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPerfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
| 376-06-7 | |
| 714.117 | |
| RUDINRUXCKIXAJ-UHFFFAOYSA-N | |
| 67822 | |
| C(=O)(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| C14HF27O2 | |
| MFCD00153263 | |
| perfluorotetradecanoic acid, perfluoromyristic acid, heptacosafluorotetradecanoic acid, tetradecanoic acid, heptacosafluoro, acmc-20ajxq, perfluorotetradecanoic acid, analytical standard | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid |
Specifications
| 376-06-7 | |
| 192°C (60 mmHg) | |
| MFCD00153263 | |
| 1811438 | |
| It is insoluble in water. | |
| C(=O)(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
| 714.117 | |
| 714.12 | |
| Perfluorotetradecanoic acid |
| 130°C to 132°C | |
| C14HF27O2 | |
| UN3261 | |
| perfluorotetradecanoic acid, perfluoromyristic acid, heptacosafluorotetradecanoic acid, tetradecanoic acid, heptacosafluoro, acmc-20ajxq, perfluorotetradecanoic acid, analytical standard | |
| RUDINRUXCKIXAJ-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid | |
| 67822 | |
| 96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 206-803-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only