missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotetradecanoic acid, 96%

Catalog No. AAL1379606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1379606 5 g
AAL1379603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1379606 Supplier Thermo Scientific Chemicals Supplier No. L1379606

CAS: 376-06-7 | C14HF27O2 | 714.117 g/mol

Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 376-06-7
Molecular Formula C14HF27O2
Molecular Weight (g/mol) 714.117
MDL Number MFCD00153263
InChI Key RUDINRUXCKIXAJ-UHFFFAOYSA-N
Synonym perfluorotetradecanoic acid, perfluoromyristic acid, heptacosafluorotetradecanoic acid, tetradecanoic acid, heptacosafluoro, acmc-20ajxq, perfluorotetradecanoic acid, analytical standard
PubChem CID 67822
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 130°C to 132°C
Boiling Point 192°C (60 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 1811438
Solubility Information It is insoluble in water.
Formula Weight 714.12
Percent Purity 96%
Chemical Name or Material Perfluorotetradecanoic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 206-803-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only