missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotetradecanoic acid, ≥96.0% (GC); 96.0-104.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111019142
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019142 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019142 Supplier Merck Emd Millipore Supplier No. 8031250MG
Only null left
Environmental

Specifications

CAS 376-06-7
Linear Formula CF3(CF2)12 CO2 H
MDL Number MFCD00153263
Melting Point 130°C to 135°C
Boiling Point 270°C (740 mmHg)
Type Analytical Standard
Quantity 50 mg
Formula Weight 714.11
Percent Purity ≥96.0% (GC); 96.0 to 104.0% (T)
Grade Analytical Standard