missing translation for 'onlineSavingsMsg'
Learn More

Perfluoroheptanes, mixed isomers, 97%, Thermo Scientific Chemicals

Catalog No. p-7046964
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1697914 25 g
AAL1697922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1697914 Supplier Thermo Scientific Chemicals Supplier No. L1697914
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 335-57-9
Molecular Formula C7F16
Molecular Weight (g/mol) 388.051
MDL Number MFCD00040339
InChI Key LGUZHRODIJCVOC-UHFFFAOYSA-N
Synonym hexadecafluoroheptane, perfluoroheptane, perfluoro-n-heptane, heptane, hexadecafluoro, perfluoroheptanes, unii-i23zvd1p1l, i23zvd1p1l, heptane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecafluoro, perfluoroheptane s, acmc-20akry
PubChem CID 9553
ChEBI CHEBI:38847
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecafluoroheptane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.72
Boiling Point 80°C to 82°C
Refractive Index 1.265
Linear Formula CF3(CF2)5CF3
Quantity 25 g
Beilstein 1716335
Formula Weight 388.05
Percent Purity 98%
Chemical Name or Material Perfluoroheptanes, mixed isomers
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-392-1
RTECSNumber MJ0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only