accessibility menu, dialog, popup

UserName

CAS RN 335-57-9

IUPAC Name: hexadecafluoroheptane
Synonyms: perfluoroheptane
Molecular Weight (g/mol): 388.05
Molecular Formula: C7F16
InChi Key: LGUZHRODIJCVOC-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)