missing translation for 'onlineSavingsMsg'
Learn More

Perfluorobutyl iodide, 99%

Catalog No. AC269950250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269950250 25 g Glass bottle
AC269951000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC269950250 Supplier Thermo Scientific Chemicals Supplier No. 269950250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 423-39-2 | C4F9I | 345.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 423-39-2
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.92
MDL Number MFCD00001062
InChI Key PGRFXXCKHGIFSV-UHFFFAOYSA-N
Synonym perfluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluorobutyliodide, n-nonafluorobutyl iodide, perfluoro-n-butyl iodide, 1-iodoperfluorobutane, ccris 9008, 1-iodononafluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo
PubChem CID 67917
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
SMILES C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F

Specifications

Melting Point -88°C
Color Purple
Density 2.0100g/mL
Boiling Point 64°C to 66°C (973.0 mbar)
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3275 to 1.3295
Linear Formula CF3(CF2)3I
Quantity 25 g
Specific Gravity 2.01
Solubility Information Solubility in water: immiscible
Formula Weight 345.92
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Perfluorobutyl iodide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 207-025-8

RUO – Research Use Only