accessibility menu, dialog, popup

UserName

FFFFFFFFFI

CAS RN 423-39-2

IUPAC Name: 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Synonyms: 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Molecular Weight (g/mol): 345.93
Molecular Formula: C4F9I
InChi Key: PGRFXXCKHGIFSV-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Boiling Point 
  • (2)
  • (6)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)