missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoro(tetradecahydrophenanthrene), mixture of isomers, Thermo Scientific Chemicals
Description
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).
Solubility
Immiscible with water.
Notes
Incompatible with lithium, sodium/sodium oxides, potassium and calcium.
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).
Solubility
Immiscible with water.
Notes
Incompatible with lithium, sodium/sodium oxides, potassium and calcium.
Specifications
Specifications
| Melting Point | -20°C |
| Density | 2.03 |
| Boiling Point | 215°C |
| Refractive Index | 1.3348 |
| Quantity | 25 g |
| Solubility Information | Immiscible with water. |
| Formula Weight | 624.12 |
| Chemical Name or Material | Perfluoro(tetradecahydrophenanthrene), mixture of isomers |
Safety and Handling
| EINECSNumber | 400-470-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only