missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(tetradecahydrophenanthrene), mixture of isomers, Thermo Scientific Chemicals

Catalog No. AAA1889822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1889822 100 g
AAA1889814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1889822 Supplier Thermo Scientific Chemicals Supplier No. A1889822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).

Solubility
Immiscible with water.

Notes
Incompatible with lithium, sodium/sodium oxides, potassium and calcium.

Chemical Identifiers

CAS 306-91-2
Molecular Formula C14F24
Molecular Weight (g/mol) 624.116
MDL Number MFCD00042596
InChI Key QKENRHXGDUPTEM-UHFFFAOYSA-N
Synonym perfluorophenanthrene, perfluoroperhydrophenanthrene, flutec pp11, perfluoro perhydrophenanthrene, tetracosafluorotetradecahydrophenanthrene, phenanthrene, tetracosafluorotetradecahydro, phenanthrene, 1,1,2,2,3,3,4,4,4a,4b,5,5,6,6,7,7,8,8,8a,9,9,10,10,10a-tetracosafluorotetradecahydro, flutec tm pp11 r, acmc-1clkp, dsstox_cid_27029
PubChem CID 78972
ChEBI CHEBI:39423
IUPAC Name 1,1,2,2,3,3,4,4,4a,4b,5,5,6,6,7,7,8,8,8a,9,9,10,10,10a-tetracosafluorophenanthrene
SMILES C12(C3(C(C(C(C1(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)(F)F)(F)F)(C(C(C(C3(F)F)(F)F)(F)F)(F)F)F)F)F

Specifications

Melting Point -20°C
Density 2.03
Boiling Point 215°C
Refractive Index 1.3348
Quantity 100 g
Solubility Information Immiscible with water.
Formula Weight 624.12
Chemical Name or Material Perfluoro(tetradecahydrophenanthrene), mixture of isomers
EINECSNumber 400-470-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only