missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(methyldecahydronaphthalene), tech. 85%, mixture of isomers, Thermo Scientific Chemicals

Catalog No. AAL0586214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0586214 25 g
AAL0586222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0586214 Supplier Thermo Scientific Chemicals Supplier No. L0586214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51294-16-7
Molecular Formula C11F20
Molecular Weight (g/mol) 512.09
MDL Number MFCD00014326
InChI Key LWRNQOBXRHWPGE-UHFFFAOYNA-N
Synonym perfluoromethyldecalin, perfluoro methyldecalin, flutec pp9, perfluoro 1-methyldecalin, perfluoro-1-methyldecalin, pp 9, perfluoro methyldecalin , tech., naphthalene, 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluorodecahydro-8-trifluoromethyl, heptadecafluorodecahydro-1-trifluoromethyl naphthalene, 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluoro-8-trifluoromethyl naphthalene
PubChem CID 39977
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluoro-8-(trifluoromethyl)naphthalene
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C12F

Specifications

Melting Point -70°C
Density 1.972
Boiling Point ∼160°C
Refractive Index 1.317
Quantity 25 g
Formula Weight 512.09
Percent Purity 85%
Grade Technical
Chemical Name or Material Perfluoro(methyldecahydronaphthalene), ≈1:1 mixture of 1- and 2-isomers
RTECSNumber QJ7175000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only