accessibility menu, dialog, popup

UserName

CAS RN 51294-16-7

IUPAC Name: 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluoro-8-(trifluoromethyl)-decahydronaphthalene
Synonyms: perfluoromethyldecalin
Molecular Weight (g/mol): 512.09
Molecular Formula: C11F20
InChi Key: LWRNQOBXRHWPGE-UHFFFAOYNA-N
SMILES: FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C12F
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)