missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals

Catalog No. AAA1828814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1828814 25 g
AAA1828822 100 g
AAA1828836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1828814 Supplier Thermo Scientific Chemicals Supplier No. A1828814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 306-94-5
Molecular Formula C10F18
Molecular Weight (g/mol) 462.081
MDL Number MFCD00010626
InChI Key UWEYRJFJVCLAGH-UHFFFAOYSA-N
Synonym perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
PubChem CID 9386
ChEBI CHEBI:38848
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
SMILES C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F

Specifications

Melting Point -10°C to -7°C
Density 1.94
Boiling Point 141°C to 142°C
Refractive Index 1.3145
Quantity 25 g
Beilstein 2067113
Merck Index 14,4183
Solubility Information Insoluble in water.
Formula Weight 462.1
Percent Purity 95%
Chemical Name or Material Perfluoro(decahydronaphthalene), cis + trans
Show More Show Less
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 206-192-4
RTECSNumber QJ3175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only