accessibility menu, dialog, popup

UserName

CAS RN 306-94-5

IUPAC Name: octadecafluoro-decahydronaphthalene
Synonyms: perfluorodecalin FDC perflunafene
Molecular Weight (g/mol): 462.08
Molecular Formula: C10F18
InChi Key: UWEYRJFJVCLAGH-UHFFFAOYSA-N
SMILES: FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F
Boiling Point 
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)