missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(2-butyltetrahydrofuran) (so called), TCI America™

Catalog No. P086725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P086725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P086725G Supplier TCI America Supplier No. P086725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 335-36-4
Molecular Formula C8F16O
Molecular Weight (g/mol) 416.06
MDL Number MFCD00014513
InChI Key FYJQJMIEZVMYSD-UHFFFAOYNA-N
Synonym perfluoro-2-butyltetrahydrofuran, perfluoro 2-butyltetrahydrofuran, fluorinert fc-75, perfluoro-compound fc-75, perfluoro butyltetrahydrofuran, heptafluorotetrahydro-2-nonafluorobutyl furan, furan, 2,2,3,3,4,4,5-heptafluorotetrahydro-5-nonafluorobutyl, perfluoro-2-n-butyltetrahydrofurane, 2,2,3,3,4,4,5-heptafluoro-5-nonafluorobutyl tetrahydrofuran, 2,2,3,3,4,4,5-heptafluorotetrahydro-5-nonafluorobutyl furan
PubChem CID 78976
ChEBI CHEBI:39023
IUPAC Name 2,2,3,3,4,4,5-heptafluoro-5-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)oxolane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)OC(F)(F)C(F)(F)C1(F)F

Specifications

Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 102°C
Formula Weight 416.06
Chemical Name or Material Perfluoro(2-butyltetrahydrofuran) (so called)
EINECSNumber (5)-0071
TSCA Yes