accessibility menu, dialog, popup

UserName

FFFFFFFFFFOFFFFFF

CAS RN 335-36-4

IUPAC Name: 2,2,3,3,4,4,5-heptafluoro-5-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)oxolane
Synonyms: 2,2,3,3,4,4,5-heptafluoro-5-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)oxolane
Molecular Weight (g/mol): 416.06
Molecular Formula: C8F16O
InChi Key: FYJQJMIEZVMYSD-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)OC(F)(F)C(F)(F)C1(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results