missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoro-1-butanesulfonyl Fluoride 96.0+%, TCI America™
Supplier: TCI America P109825G
Specifications
| Perfluoro-1-butanesulfonyl Fluoride | |
| Yellow | |
| C4F10O2S | |
| 25 g | |
| nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro | |
| C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F | |
| 302.086 | |
| 302.09 | |
| Liquid |
| 375-72-4 | |
| 65°C | |
| MFCD00007422 | |
| 3265 | |
| LUYQYZLEHLTPBH-UHFFFAOYSA-N | |
| 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride | |
| 67814 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 375-72-4 | |
| 302.086 | |
| LUYQYZLEHLTPBH-UHFFFAOYSA-N | |
| 67814 | |
| C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F |
| C4F10O2S | |
| MFCD00007422 | |
| nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro | |
| 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride |
Safety and Handling
TSCA : Yes