accessibility menu, dialog, popup

UserName

CAS RN 375-72-4

IUPAC Name: nonafluorobutane-1-sulfonyl fluoride
Synonyms: perfluorobutanesulfonyl fluoride
Molecular Weight (g/mol): 302.09
Molecular Formula: C4F10O2S
InChi Key: LUYQYZLEHLTPBH-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(F)(=O)=O
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)