missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-butanesulfonyl fluoride, 92%

Catalog No. p-179856
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312340050 5 g Glass bottle
AC312340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312340050 Supplier Thermo Scientific Chemicals Supplier No. 312340050
Only null left

CAS: 375-72-4 | C4F10O2S | 302.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 375-72-4
Molecular Formula C4F10O2S
Molecular Weight (g/mol) 302.09
MDL Number MFCD00007422
InChI Key LUYQYZLEHLTPBH-UHFFFAOYSA-N
Synonym nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro
PubChem CID 67814
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride
SMILES C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F

Specifications

Color Colorless to Yellow
Density 1.6800g/mL
Boiling Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)3SO2F
Refractive Index 1.3
Quantity 5 g
Specific Gravity 1.68
Formula Weight 302.09
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Perfluoro-1-butanesulfonyl fluoride, 92%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-792-6
TSCA TSCA

RUO – Research Use Only