missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech., Thermo Scientific Chemicals

Catalog No. p-7025813
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2150018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2150018 Supplier Thermo Scientific Chemicals Supplier No. B2150018

It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 374-76-5
Molecular Formula C9F18
Molecular Weight (g/mol) 450.07
MDL Number MFCD00066614
InChI Key MGOFOLPXKULBGG-UHFFFAOYSA-N
Synonym perfluoro-1,3,5-trimethylcyclohexane, 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl cyclohexane, acmc-1ajc4, nonafluoro-1,3,5-tris-trifluoromethyl-cyclohexane, 1,2,2,3,4,4,5,6,6-nonafluoro-1,3,5-tris trifluoromethyl cyclohexane, cyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl, 1,1,2,3,3,4,5,5,6-nonakis fluoranyl-2,4,6-tris trifluoromethyl cyclohexane
PubChem CID 2776392
IUPAC Name 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Melting Point -68°C
Density 1.888
Boiling Point 125°C to 128°C
Refractive Index 1.297
Quantity 50 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 450.07
Grade Technical
Chemical Name or Material Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only