accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFFFF

CAS RN 374-76-5

IUPAC Name: 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
Synonyms: 1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
Molecular Weight (g/mol): 450.07
Molecular Formula: C9F18
InChi Key: MGOFOLPXKULBGG-UHFFFAOYSA-N
SMILES: FC(F)(F)C1(F)C(F)(F)C(F)(C(F)(F)F)C(F)(F)C(F)(C(F)(F)F)C1(F)F
Boiling Point 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)