Learn More
Perfex™ TruCLEAN™ Buckets and Pails
$70.05 - $205.49
Specifications
Products 7
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19-130-3423
View Documents
Perfex™
2215B
Each for $103.57
Only null left
19-003410R
View Documents
Perfex™
PER2215R
Each for $110.98
Only null left
19-003410W
View Documents
Perfex™
PER2215W
Each for $110.98
Only null left
19-0034200B
View Documents
Perfex™
PER2206B
Each for $70.05
Only null left
19-0034200R
View Documents
Perfex™
PER2206R
Each for $70.05
Only null left
19-0034200W
View Documents
Perfex™
PER2206W
Each for $70.05
Only null left
19-0034201
View Documents
Perfex™
PER2251
Each for $205.49
Only null left
Description
Description
For use with TruCLEAN Pro System; Also available: cart handle pail basket.
Buckets and pails feature nonporous, super smooth surface that prevents microbial growth; embossed graduations in English and metric; stainless-steel handles. Compatible with gamma, EtO, and autoclave sterilization. 4 gal. (15L) buckets fit TruCLEAN Pro Triple Bucket System cart. Two 1.6 gal. (6L) pails can fit into the stainless-steel basket that rests on TruCLEAN Pro cart handle. Buckets and pails are available in blue, red, or white.
Specifications
Specifications
0.0