Learn More
Perfex™ TruCLEAN™ Buckets and Pails
Supplier:  Perfex™ PER 2206B
 View more versions of this product
Catalog No. 190034200B
Only null left
Description
Description
For use with TruCLEAN Pro System; Also available: cart handle pail basket.
Buckets and pails feature nonporous, super smooth surface that prevents microbial growth; embossed graduations in English and metric; stainless-steel handles. Compatible with gamma, EtO, and autoclave sterilization. 4 gal. (15L) buckets fit TruCLEAN Pro Triple Bucket System cart. Two 1.6 gal. (6L) pails can fit into the stainless-steel basket that rests on TruCLEAN Pro cart handle. Buckets and pails are available in blue, red, or white.
Product Suggestions
Product Suggestions