Learn More
Requires : Requires 19-042-036 or 19-042-041 handle.
Description
Provide exceptional scrubbing action on irregular surfaces as well as floors, walls, and ceilings with these moderately stiff poly brushes. Nonconductive polyester fibers are flared to maintain contact at any angle, even on uneven surfaces. Unlike nylon, the polyester fibers and block will not absorb bacteria, liquids, or odors. Ergonomically designed to eliminate awkward bending and reaching. Available in blue, white, and red.
Chemical Identifiers
| Multi-purpose Brush | |
| Provides exceptional scrubbing action on a variety of irregular surfaces, as well as floors, walls and ceilings | |
| Adjustable Polymer Handle | |
| Polyester Fiber |
Specifications
| Multi-purpose Brush | |
| Provides exceptional scrubbing action on a variety of irregular surfaces, as well as floors, walls and ceilings | |
| Polyester Fiber |
| Red | |
| Adjustable Polymer Handle |