missing translation for 'onlineSavingsMsg'
Learn More

Perfex™ Multipurpose Brushes

Catalog No. p-25895
Change view
Click to view available options
Color:
Blue
Red
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19042034A Blue
19042034B Red
19042034C White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19042034A Supplier Perfex™ Supplier No. 3040B
Only null left

Requires

Requires 19-042-036 or 19-042-041 handle.

Provide exceptional scrubbing action on irregular surfaces as well as floors, walls, and ceilings with these moderately stiff poly brushes. Nonconductive polyester fibers are flared to maintain contact at any angle, even on uneven surfaces. Unlike nylon, the polyester fibers and block will not absorb bacteria, liquids, or odors. Ergonomically designed to eliminate awkward bending and reaching. Available in blue, white, and red.

Specifications

Product Type Multi-purpose Brush
Color Blue
For Use With (Application) Provides exceptional scrubbing action on a variety of irregular surfaces, as well as floors, walls and ceilings
For Use With (Equipment) Adjustable Polymer Handle
Material Polyester Fiber