missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Perfektum™ Rustless Steel Syringe Needles
Robust stainless-steel design offers accurate, reproducible injections
Supplier: Cadence Science™ POP4047
Description
- Chrome-plated brass female luer hub
- Corrosion-resistant Type 304 stainless steel needle
- Regular medical point
- Variety of sizes and gauges available
- Approved for laboratory use only
Specifications
| Chrome-plated Brass (Hub), Stainless Steel | |
| For laboratory use only | |
| Hypodermic Needle | |
| 76 mm | |
| Female Luer |
| 22 G | |
| 3 in. | |
| 12 Needles | |
| Regular Medical | |
| Non-sterile |