missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Perfect Fit 13G GY PU A5/E Cut-resistant Gloves

The Honeywell Perfect Fit gloves are engineered to be lightweight, glass fiber-free, and coated with a specialty engineered Polyurethane (PU); this PU coating is designed with a low level of dimethylformamide (DMF) which protects your workers at a higher level.

$15.01 - $15.01

Specifications

Coating Material Polyurethane (DMF)
Color Gray
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free No
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17004346
View Documents
Honeywell™
NPF259113G6XS
6, X-Small
Pair for $15.01
Pack of 10 Pair for $143.30
Only null left
 
17004347
View Documents
Honeywell™
NPF259113G7S
7, Small
Pair for $15.01
Pack of 10 Pair for $143.30
Only null left
 
17004348
View Documents
Honeywell™
NPF259113G8M
8, Medium
Pair for $15.01
Pack of 10 Pair for $143.30
Only null left
 
17004349
View Documents
Honeywell™
NPF259113G9L
9, Large
Pair for $15.01
Pack of 10 Pair for $143.30
Only null left
 
17004350
View Documents
Honeywell™
NPF259113G10XL
10, X-Large
Pair for $15.01
Pack of 10 Pair for $143.30
Only null left
 
17004351
View Documents
Honeywell™
NPF259113G11XXL
11, 2X-Large
Pair for $15.01
Pack of 10 Pair for $143.30
Only null left
 
Description

Description

We engineered the Honeywell Perfect Fit Gloves using advanced technologies and unique features. The gloves are designed to help workers perform at their best with enhanced comfort, dexterity, grip and protection.

INDUSTRIES:

Automotive, Machinery and Equipment, Metal Fabrication , Warehousing and Logistics, Manufacturing, Construction, Transportation

APPLICATIONS:

Handling sharp objects, Painting and surface inspections, Chassis assembly, Mounting and dismantling, Maintenance, Fastening components and parts

SECIFICATIONS:

  • gray Perfect Fit 13G GY PU A5/E
  • Dry Grip
  • Mechanical, puncture (4), and abrasion (4) protection
  • Food certified
  • Polyurethane coating
  • Cut Protection
  • Tear resistant
  • Pack of 10
  • Size range: 6 to 11
  • Silicone Free
  • PPE Category: 2
  • Cuff Length: 300 inch
  • Wrist type: Extended Wrist
  • Latex free: No
  • Cuff Sytle: Wristband
  • Standards: ANSI/ISEA 105:2016, EN ISO 21420, EN 388, ANSI/ISEA 105, EU 1935/2004
  • Washable: Y
  • Material: Polyamide, polyurethane
  • Color: gray
Specifications

Specifications

Polyurethane (DMF)
ANSI A5
No
Gray
Specific
Cut-resistant Gloves
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'