missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide solution, PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur., Honeywell Fluka™

Catalog No. p-7072042
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000807 1 L
6000809 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000807 Supplier Honeywell Inc Supplier No. 316421L

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur.

Honeywell ACS solvents for general laboratory use are:

  • a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested tmeet ACS general use specifications

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

UN Number 2014
Beilstein 3587191
Concentration 30% (w/w)
Formula Weight 34.01g/mol
Grade puriss. p.a.
Vapor Pressure 23.3mmHg (30°C)
Quantity 1 L
Chemical Name or Material Hydrogen Peroxide Solution, Reag. Ph. Eur. reag. ISO
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H318-H412