missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, extra pure, ca. 70% solution in water

Catalog No. p-4813596
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452850010 1 L
AC451770025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452850010 Supplier Thermo Scientific Chemicals Supplier No. 452850010
Only null left

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7601-90-3 , 7732-18-5
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point -18.0°C
Boiling Point 203.0°C
Density 1.6600g/mL
Specific Gravity 1.66
Flash Point 113°C
Quantity 1 L
Viscosity 3.5 mPa.s (20°C)
Linear Formula HClO4
Solubility Information Solubility in water: soluble.
Formula Weight 100.46
Grade Analytical
Chemical Name or Material Perchloric Acid
Show More Show Less

RUO – Research Use Only