missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid Solution, N/10 (0.1N) (Certified), Fisher Chemical™

Catalog No. p-20418
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP339500 500 mL Glass Bottle
SP3394 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP339500 Supplier Thermo Fisher Scientific Supplier No. SP339 500
Only null left

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 7601-90-3 , 64-19-7
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Concentration 0.0995 to 0.1005N
Density 1.060g/cm³
Packaging Glass Bottle
Traceability to NIST Lot Number Used (KHP Standard)
Quantity 500 mL
Physical Form Liquid
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N (N/10)
Chemical Name or Material Perchloric Acid Solution, N/10 (0.1N)
Show More Show Less
DOTInformation DOT Class 8, : Corrosive