missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, for analysis, ca. 70% solution in water

Catalog No. p-21205
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223312500 250 mL Glass bottle
AC223310010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223312500 Supplier Thermo Scientific Chemicals Supplier No. 223312500
Only null left

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent for analysis

Chemical Identifiers

CAS 7601-90-3 , 7732-18-5
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point -18.0°C
Boiling Point 203.0°C
Density 1.6600g/mL
Heavy Metals (as Pb) 1ppm max.
Packaging Glass bottle
Specific Gravity 1.66
Flash Point 113°C
Quantity 250 mL
Viscosity 3.5 mPa.s (20°C)
Physical Form Liquid
Linear Formula HClO4
Merck Index 15, 7267
Solubility Information Solubility in water: soluble.
Formula Weight 100.46
Percent Purity 70 to 72%
Grade Analytical
Chemical Name or Material Perchloric acid, p.a.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-512-4
RTECSNumber SC7500000
TSCA TSCA

RUO – Research Use Only