Learn More
Perchloric acid, for analysis, ca. 70% solution in water
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Supplier: Thermo Scientific Chemicals 223310010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysisSpecifications
| Perchloric acid | |
| -18.0°C | |
| 203.0°C | |
| 1ppm max. | |
| 1.66 | |
| 1 L | |
| ClHO4 | |
| HClO4 | |
| 15, 7267 | |
| Solubility in water: soluble. | |
| O[Cl](=O)(=O)=O | |
| 100.45 | |
| CHEBI:29221 | |
| 70 to 72% |
| p.a. | |
| 7601-90-3,7732-18-5 | |
| 1.6600g/mL | |
| Glass bottle | |
| 113°C | |
| 3.5 mPa.s (20°C) | |
| Liquid | |
| MFCD00011325 | |
| unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
| VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
| perchloric acid | |
| 24247 | |
| 100.46 | |
| Analytical |
Chemical Identifiers
| 7601-90-3 | |
| 100.45 | |
| VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
| 24247 | |
| perchloric acid |
| ClHO4 | |
| MFCD00011325 | |
| unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4 | |
| CHEBI:29221 | |
| O[Cl](=O)(=O)=O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 231-512-4
RTECSNumber : SC7500000
TSCA : TSCA
RUO – Research Use Only