missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 8% (w/v) Aqueous, Ricca Chemical

Supplier:  Ricca Chemical Company R5512800100A

Catalog No. R5512800100

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Perchloric Acid
7732-18-5,7601-90-3
8% w/v
Natural Poly Bottle
100 mL
Liquid
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
Laboratory
Colorless
88.0,9.97
92.0,10.38
Aqueous Solution
ClHO4
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
8% W/V

Chemical Identifiers

7732-18-5
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions