missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 2.00 Normal, Aqueous, Reagents

Catalog No. 01334290
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334290 1 L
01334289 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334290 Supplier Labchem Inc Supplier No. CP1213001C
Only null left

Perchloric Acid, 2.00 Normal, Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7601-90-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 101°C
Packaging Glass Bottle
Specific Gravity 1.1145
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 2.00 Normal, Aqueous